missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette tips
GREENER_CHOICE

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.

Brand:  Thermo Scientific™ 2770

89.67 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10399360

  • 121.00 € / Pack of 960
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to Aid in Ppipetteting for a Variety of Applications:

Extended length: to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading: for loading acrylamide or agarose gels

Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials

Solvent safe: for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ART 200U, Filtered, Sterile, Lift-off Lid Rack
Filtered
Not Autoclavable
2.06 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Ergonomic
ART
200 μL
Sterile
ART 200U
5.2cm
Lift-off Lid Rack
Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus
Sterile & PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers