Learn More
Thermo Scientific™ ART™ SolventSafe Pipette Tips
Perform applications accurately and consistently with these Solventsafe pipette tips, which seal when exposed to potential contaminants.
Brand: Thermo Scientific™ 5469
78.47 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ SolventSafe Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
- Sterile tips are certified free from RNase, DNase and pyrogen
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
ART SolventSafe 200, Filtered, Sterile, Rack | |
Filtered | |
ART SolventSafe 200 | |
5.88cm | |
Micro Point | |
ART |
200 μL | |
Sterile | |
2.32 in. | |
Rack | |
Universal Fit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.