missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21449-1-AP-150UL
This item is not returnable.
View return policy
Specifications
ASB6 | |
Polyclonal | |
Unconjugated | |
ASB6 | |
ASB 6, ASB6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140459 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NWX5 | |
ASB6 | |
ASB6 Fusion Protein Ag13863 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |