missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | ASB6 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16831045
|
Proteintech
21449-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16821045
|
Proteintech
21449-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
ASB6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NWX5 | |
140459 | |
ASB6 Fusion Protein Ag13863 | |
Primary | |
-20°C | |
ASB6 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ASB 6, ASB6 | |
ASB6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |