Learn More
ASB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21449-1-AP-20UL
 View more versions of this product
Product Code. 16831045
  • 145.00 € / 20µL
Explore more special offers
Specifications
ASB6
Polyclonal
Unconjugated
ASB6
ASB 6, ASB6
Rabbit
Antigen Affinity Chromatography
RUO
140459
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NWX5
ASB6
ASB6 Fusion Protein Ag13863
20 μL