Learn More
Novus Biologicals™ Asparagine synthetase Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand:  Novus Biologicals™ NBP2-55125PEP
Product Code. 18256954
  • 259.00 € / 100µL
Explore more special offers
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ASNS. The Asparagine synthetase Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
440
asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein
ASNS
100 μL
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54142. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
>80% by SDS-PAGE and Coomassie blue staining
Store at -20°C. Avoid freeze-thaw cycles.
32 kDa
E. coli
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novus Biologicals™ Asparagine synthetase Recombinant Protein Antigen > 100 μL