missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain AstiPure™ RF1 Series PFA Male Tube Connector

Use these fittings to deliver corrosive fluids. Saint Gobain Performance Plastics™ AstiPure™ RF1 Series PFA Male Tube Connector are leak proof and reliable under a wide range of temperatures and pressures.

Brand:  Saint-Gobain RF10410

 View more versions of this product

Product Code. 11974139

  • 84.10 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Sturdy rubber Silicone piston rings for flasks
  • For general lab applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PFA
For 4 x 6mm dia. tubing
12mm
Male Tube Connector
40 mm
1 Each
1/8 in. NPT
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Saint-Gobain AstiPure™ RF1 Series PFA Male Tube Connector > Length: 40mm; Thread Size: 1/8 in. NPT

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.