Learn More
ASTM D-6584 Mix 3 (4C) Standard Solution, Chem Lab
Brand: Chem Lab CL40.13783.0001
Specifications
Ampoule in glass | |
0.98 g/mL | |
Liquid | |
-42°C | |
79.1 |
1 mL | |
17 °C | |
Colorless | |
115°C | |
Pyridine |
Safety and Handling
- ASTM D-6584 Mix 3 (4C) standard solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
- MIXTURE LIST-Contains : ASTM
Your input is important to us. Please complete this form to provide feedback related to the content on this product.