Learn More
Fisherbrand™ Model 50 Sonic Dismembrator
Economical, small volume cell disruptor.
Brand: Fisherbrand™ FB50EUK-220
Includes: Generator, converter, 1/8 in. probe, cables, wrench set
Description
The Fisher Scientific™ Model 50 Sonic Dismembrator is compact, portable and extremely simple to operate. Weighing less than 4 lb., this model is the smallest unit on the market and is highly effective for cell disruption, sample preparation and many other small volume applications.
- Processes 0.2 to 50mL
- 50 watts
- Continuous or Remote operation (footswitch optional)
- Available accessories include: 3 Probe sizes for individual samples
- Lightweight converter for handheld operation
- Basic small volume applications
Order Info
Stand not included.
Services
Two years
Specifications
220/230 V | |
20 kHz (Ultrasonic Frequency) | |
0.2 to 50 mL | |
5 in. | |
7 in. | |
8 in. | |
Ideal for basic cell disruption | |
FB50 |
220/230 V, 50/60 Hz | |
50 W | |
7 x 8 x 5 in. (17.78 x 20.32 x 12.7 cm) | |
12.7 cm | |
17.78 cm | |
20.32 cm | |
Generator, converter,1/8 in. dia. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.