Learn More
MedChemExpress ATP-polyamine-biotin
MedChemExpress ATP-polyamine-biotin, the first cell-permeable ATP analogue, is an efficient kinase cosubstrate. ATP-polyamine-biotin promotes biotin labeling of kinase substrates in live cells.
716.85 € - 2231.55 €
Specifications
Product Type | Biochemical Assay Reagents |
---|---|
Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
Shipping Condition | Room temperature |
CAS | 1800401-93-7 |
Molecular Weight (g/mol) | 945.85 g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16492409
|
MedChemExpress
HY-D0183 |
1 mg |
716.85 €
1mg |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||||
16490376
|
MedChemExpress
HY-D0183 |
100 mg |
1275.75 €
2mg |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
16421168
|
MedChemExpress
HY-D0183 |
5 mg |
2231.55 €
5mg |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
Specifications
Biochemical Assay Reagents | |
Room temperature | |
945.85 g/mol | |
HBV |
4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) | |
1800401-93-7 | |
H2O : 6 mg/mL (6.34 mM; Need ultrasonic and warming) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.