missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene T-Connector Barbed Fittings

These Barbed T Connectors are designed for quick and easy assembly in low-pressure applications.

Brand:  Masterflex™ 40623-70

 View more versions of this product

Product Code. 15246858

  • 84.50 € / Pack of 25
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simple and economical
  • For use with flexible tubing
  • Connects fluid paths in the laboratory
  • Creates secure seals that typically hold pressures up to 30 psi (2 bar)
  • No leaks, flow restrictions or costly downtime
Specifications

Specifications

Barbed Fitting
Polypropylene
-13°C
Autoclave or Ethylene Oxide
Clear
Autoclavable
3/16 in. tubing I.D.
25 Pack
T-shape
66°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.