Learn More
Thermo Scientific™ Automation Reservoirs, with removable splash baffles
Handle reagents on automated systems simply and economically.
232.00 € - 335.00 €
Product Code | Brand | Channels | Sterility | Description | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Channels | Sterility | Description | Material | Price | Quantity & Availability | |||
10416343
![]() |
Thermo Scientific™
1064-15-8 |
96 | Sterile | Polypropylene |
260.00 €
Pack of 20 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10253343
![]() |
Thermo Scientific™
1064-15-6 |
96 | Sterile | Deepwell | Polypropylene |
335.00 €
Pack of 20 |
In Stock
Log in to see stock. |
|||
10141443
![]() |
Thermo Scientific™
1064-15-7 |
384 | Sterile | Polypropylene |
260.00 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
10795394
![]() |
Thermo Scientific™
1064-05-7 |
384 | Non-sterile | Polypropylene |
232.00 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
10131443
![]() |
Thermo Scientific™
1064-05-8 |
96 | Non-sterile | Polypropylene |
249.00 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
10101443
![]() |
Thermo Scientific™
1064-05-6 |
96 | Non-sterile | Deepwell | Plastic |
299.00 €
Pack of 20 |
In Stock
Log in to see stock. |
|||
Description
Thermo Scientific™ Automation Reservoirs provide standard SBS dimensions ensuring compatibility with plate stackers and handlers, which enables you to easily handle reagents on automated systems economically. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Features:- Standard microplate dimensions enables reservoirs to be used in plate handlers or stackers
- Hydrophilic surface so liquid spreads out evenly with minimal reagent needed to cover bottom
- Polypropylene construction offers excellent chemical resistance
- Available individually wrapped for sterile use
Removable Splash Baffles:
- Prevent reagents from splashing out when being moved on robotic decks
- Enable reservoirs to be moved faster so they can also be filled closer to the top with reagent
- Can be removed to increase reservoirs capacity
Specifications
Reservoir | |
Removable splash baffles |