Learn More
Thermo Scientific™ Automation Reservoirs, with removable splash baffles
GREENER_CHOICE
Handle reagents on automated systems simply and economically.
Brand:  Thermo Scientific™ 1064-15-8
188.00 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 10416343
  • 260.00 € / Pack of 20
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Automation Reservoirs provide standard SBS dimensions ensuring compatibility with plate stackers and handlers, which enables you to easily handle reagents on automated systems economically. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Features:
  • Standard microplate dimensions enables reservoirs to be used in plate handlers or stackers
  • Hydrophilic surface so liquid spreads out evenly with minimal reagent needed to cover bottom
  • Polypropylene construction offers excellent chemical resistance
  • Available individually wrapped for sterile use
Removable Splash Baffles:
  • Prevent reagents from splashing out when being moved on robotic decks
  • Enable reservoirs to be moved faster so they can also be filled closer to the top with reagent
  • Can be removed to increase reservoirs capacity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
Removable splash baffles
96
Polypropylene
Reservoir
Individually Wrapped
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Automation Reservoirs, with removable splash baffles > Automation Reservoirs; Format: 96 channel; Sterility: Sterile; Packaging: Individually wrapped; Unit Size: Case of 20 pcs