Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Brand:  Thermo Scientific™ 921-021
449.97 EUR valid until 2025-12-31
Use promo code "24289" to get your promotional price.
 View more versions of this product
Product Code. 11750095
  • 692.00 € / Pack of 4800
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
TRUSTED_SUSTAINABILITY
Specifications
Specifications
80 μL
Sterile
Rack
Zymark/Caliper Liquid Handling Workstations
Sterile and PCR Ready
Filtered
Clear
Automation
Caliper Zymark
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations > 80uL; Sterile; ART filter; Low retention; 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis