missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

374.00 € - 692.00 €

Specifications

Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
View More Specs

Products 8
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
16406949
GREENER_CHOICE
View Documents
Thermo Scientific™
920-021
50 μL Filtered Sterile Sterile and PCR Ready
692.00 €
Pack of 4800
Estimated Shipment: 13-08-2025
to see stock.
Add to basket
 
16416949
GREENER_CHOICE
View Documents
Thermo Scientific™
920-261
50 μL Non-filtered Sterile Sterile
398.00 €
Pack of 4800
Estimated Shipment: 13-08-2025
to see stock.
Add to basket
 
16426949
GREENER_CHOICE
View Documents
Thermo Scientific™
920-262
50 μL Non-filtered Non-sterile Certified Quality
374.00 €
Pack of 4800
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
 
11750095
GREENER_CHOICE
View Documents
Thermo Scientific™
921-021
80 μL Filtered Sterile Sterile and PCR Ready
692.00 €
Pack of 4800
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
 
11860421
GREENER_CHOICE
View Documents
Thermo Scientific™
921-262
100 μL Non-filtered Non-sterile Certified Quality
374.00 €
Pack of 4800
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
 
11760095
GREENER_CHOICE
View Documents
Thermo Scientific™
923-021
150 μL Filtered Sterile Sterile and PCR Ready
692.00 €
Pack of 4800
In Stock
to see stock.
Add to basket
 
16436949
GREENER_CHOICE
View Documents
Thermo Scientific™
923-261
200 μL Non-filtered Sterile Sterile
398.00 €
Pack of 4800
Estimated Shipment: 13-08-2025
to see stock.
Add to basket
 
11806081
GREENER_CHOICE
View Documents
Thermo Scientific™
923-262
200 μL Non-filtered Non-sterile Certified Quality
374.00 €
Pack of 4800
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
 
Description

Description

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications

Specifications

Clear
Automation
Caliper Zymark
Rack
Zymark/Caliper Liquid Handling Workstations
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.