Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Brand:  Thermo Scientific™ 903-261
128.25 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
 View more versions of this product
Product Code. 10060511
  • 135.00 € / Pack of 960
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Automation
960 Tips
Sterile
Non-filtered
Clear
ISO 9001 and ISO 13485
Rack
Reload Insert, 96 tips/insert, 10 inserts/pack
Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms > Automation tips for Tecan™ Volume: 200μL; Sterility: Sterile; Color: Clear; Unit Size: 10 blisters of 96 tips (960 tips in total)