Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
123.00 € - 1155.00 €
Specifications
Certifications/Compliance | ISO 9001 and ISO 13485 |
---|---|
Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
Format | Rack |
Product Code | Brand | Volume (Metric) | Filtered | For Use With (Equipment) | Fit for Purpose | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | For Use With (Equipment) | Fit for Purpose | Price | Quantity & Availability | |||
10181471
![]() |
Thermo Scientific™
901-021 |
10 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
176.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10338890
![]() |
Thermo Scientific™
901-251 |
20 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
176.00 €
Pack of 960 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||
10013660
![]() |
Thermo Scientific™
901-252 |
20 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
159.00 €
Pack of 960 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||
10623482
![]() |
Thermo Scientific™
901-253 |
20 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
627.00 €
Pack of 4800 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
10461093
![]() |
Thermo Scientific™
901-261 |
20 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
139.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10000481
![]() |
Thermo Scientific™
901-262 |
20 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
129.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10697831
![]() |
Thermo Scientific™
902-011 |
50 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
214.00 €
Pack of 960 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||
10115430
![]() |
Thermo Scientific™
902-251 |
50 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
176.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10740981
![]() |
Thermo Scientific™
902-252 |
50 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
159.00 €
Pack of 960 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||
10633482
![]() |
Thermo Scientific™
902-253 |
50 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
627.00 €
Pack of 4800 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10663242
![]() |
Thermo Scientific™
902-261 |
50 μL | Non-filtered | Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
139.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10673242
![]() |
Thermo Scientific™
902-262 |
50 μL | Non-filtered | Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
129.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10125430
![]() |
Thermo Scientific™
903-011 |
175 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
209.00 €
Pack of 960 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||
10562162
![]() |
Thermo Scientific™
903-011G |
175 μL | Filtered | Cavro, MiniPrep | Sterile and PCR Ready |
209.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10141571
![]() |
Thermo Scientific™
903-021 |
175 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
165.00 €
Pack of 960 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||
11510145
![]() |
Thermo Scientific™
903-033 |
175 μL | Filtered | Tecan Freedom EVO LiHa Head, Tecan Genesis LiHa Head | PCR Ready |
1155.00 €
Pack of 4800 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10439970
![]() |
Thermo Scientific™
903-251 |
200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
165.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10561972
![]() |
Thermo Scientific™
903-251G |
200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
165.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10648221
![]() |
Thermo Scientific™
903-252 |
200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
152.00 €
Pack of 960 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
|||
10772563
![]() |
Thermo Scientific™
903-252G |
200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
152.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10102821
![]() |
Thermo Scientific™
903-253 |
200 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
627.00 €
Pack of 4800 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
10060511
![]() |
Thermo Scientific™
903-261 |
200 μL | Non-filtered | Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
135.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10759221
![]() |
Thermo Scientific™
903-262 |
200 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
123.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10658221
![]() |
Thermo Scientific™
904-011 |
1000 μL | Filtered | Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
209.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10702953
![]() |
Thermo Scientific™
904-011G |
1000 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
209.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
16446939
![]() |
Thermo Scientific™
904-013 |
1000 μL | Filtered | Tecan Freedom EVO LiHa Head, Tecan Genesis | PCR Ready |
793.00 €
Pack of 4800 |
In Stock
Log in to see stock. |
|||
10345981
![]() |
Thermo Scientific™
904-021 |
1000 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
165.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10658411
![]() |
Thermo Scientific™
904-251 |
1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
165.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10231482
![]() |
Thermo Scientific™
904-251G |
1000 μL | Non-filtered | Cavro, Tecan Genesis, Tecan Freedom EVO, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
165.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10348980
![]() |
Thermo Scientific™
904-252 |
1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
152.00 €
Pack of 960 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||
10572162
![]() |
Thermo Scientific™
904-252G |
1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
153.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10565051
![]() |
Thermo Scientific™
904-253 |
1000 μL | Non-filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
627.00 €
Pack of 4800 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
10306031
![]() |
Thermo Scientific™
904-261 |
1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Sterile |
163.00 €
Pack of 960 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||
10729601
![]() |
Thermo Scientific™
904-262 |
1000 μL | Non-filtered | Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus | Certified Quality |
150.00 €
Pack of 960 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||
10073620
![]() |
Thermo Scientific™
901-011 |
10 μL | Filtered | Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro | Sterile and PCR Ready |
215.00 €
Pack of 960 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
Features:- Available as sterile, filtered and/or liquid sensing.
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
- Lot-tested during production on actual workstation for which they are created.
- Low percentage coefficient of variation (%CV).
Specifications
ISO 9001 and ISO 13485 | |
Free of RNase, DNase, DNA, ATP, and endotoxin | |
Rack |