Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand:  Thermo Scientific™ 906-262
402.19 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
 View more versions of this product
Product Code. 10325104
  • 467.00 € / Pack of 4800
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
  • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
  • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Non-sterile
Rack
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius
Certified Quality
Non-filtered
Clear
Low Retention, Extended Length
Tecan
Product Suggestions
Product Suggestions
Videos
FDS
Documents
Documents
Certificats
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm > 50μL; Non-sterile; For use with Tecan Freedom Evo, 10 Racks/Pack, 5 Packs/Case
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis