Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
467.00 € - 743.00 €
Specifications
Color Clear
Format Rack
Product Line Tecan
View More Specs
Products 9
Product Code Brand Volume (Metric) Sterility Filtered Fit for Purpose Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Filtered Fit for Purpose Price Quantity & Availability  
11897313
GREENER_CHOICE
View Documents
Thermo Scientific™
906-021
50 μL Sterile Filtered Sterile and PCR Ready
743.00 €
Pack of 4800
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
10078172
GREENER_CHOICE
View Documents
Thermo Scientific™
906-261
50 μL Sterile Non-filtered Sterile
479.00 €
Pack of 4800
Estimated Shipment: 26-08-2025
to see stock.
Add to basket
10325104
GREENER_CHOICE
View Documents
Thermo Scientific™
906-262
50 μL Non-sterile Non-filtered Certified Quality
467.00 €
Pack of 4800
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
11807323
GREENER_CHOICE
View Documents
Thermo Scientific™
907-021
80 μL Sterile Filtered Sterile and PCR Ready
743.00 €
Pack of 4800
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
10101804
GREENER_CHOICE
View Documents
Thermo Scientific™
907-261
100 μL Sterile Non-filtered Sterile
479.00 €
Pack of 4800
Estimated Shipment: 26-08-2025
to see stock.
Add to basket
10654844
GREENER_CHOICE
View Documents
Thermo Scientific™
907-262
100 μL Non-sterile Non-filtered Certified Quality
467.00 €
Pack of 4800
Estimated Shipment: 19-08-2025
to see stock.
Add to basket
11815953
GREENER_CHOICE
View Documents
Thermo Scientific™
908-021
150 μL Sterile Filtered Sterile and PCR Ready
743.00 €
Pack of 4800
In Stock
to see stock.
Add to basket
10753675
GREENER_CHOICE
View Documents
Thermo Scientific™
908-261
200 μL Sterile Non-filtered Sterile
479.00 €
Pack of 4800
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
10373653
GREENER_CHOICE
View Documents
Thermo Scientific™
908-262
200 μL Non-sterile Non-filtered Certified Quality
467.00 €
Pack of 4800
Estimated Shipment: 19-08-2025
to see stock.
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Specifications
Clear
Rack
Tecan
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm