missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
158.32 € - 2054.38 €
Specifications
Certifications/Compliance | RNase, DNase, pyrogen-free |
---|---|
Closure Material | Polypropylene |
Capacity (Metric) | 1.5 mL |
Material | Polypropylene |
Format | Non-looped |
Product Code | Brand | Sterility | Closure Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Closure Color | Price | Quantity & Availability | ||||||
11340385
|
Axygen™
SCT-150-SS-A |
Non-sterile | Assorted | N/A | N/A | N/A | |||||
12779538
|
Axygen™
SCT-150-SS-A-S |
Sterile | Assorted |
2054.38 €
Pack of 4000 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11350385
|
Axygen™
SCT-150-SS-A-S |
Sterile | Assorted | N/A | N/A | N/A | |||||
11366044
|
Axygen™
SCT-150-SS-C |
Non-sterile | Clear |
160.49 €
Pack of 500 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
13107464
|
Axygen™
SCT-150-SS-C |
Non-sterile | Clear | N/A | N/A | N/A | |||||
12719548
|
Axygen™
SCT-150-SS-C-S |
Sterile | Clear | N/A | N/A | N/A | |||||
12799538
|
Axygen™
SCT-150-SS-BR |
Non-sterile | Brown | N/A | N/A | N/A | |||||
13124624
|
Axygen™
SCT-150-SS-G |
Non-sterile | Green | N/A | N/A | N/A | |||||
12739548
|
Axygen™
SCT-150-SS-GR |
Non-sterile | Gray | N/A | N/A | N/A | |||||
12789548
|
Axygen™
SCT-150-SS-O |
Non-sterile | Orange | N/A | N/A | N/A | |||||
12799548
|
Axygen™
SCT-150-SS-P |
Non-sterile | Pink | N/A | N/A | N/A | |||||
12709558
|
Axygen™
SCT-150-SS-R |
Non-sterile | Red | N/A | N/A | N/A | |||||
12719558
|
Axygen™
SCT-150-SS-V |
Non-sterile | Violet | N/A | N/A | N/A | |||||
12729558
|
Axygen™
SCT-150-SS-W |
Non-sterile | White | N/A | N/A | N/A | |||||
12749558
|
Axygen™
SCT-150-SS-Y |
Non-sterile | Yellow | N/A | N/A | N/A | |||||
Description
For collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Pre-sterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order
- Caps available in range of colors for easy sample identification
Specifications
RNase, DNase, pyrogen-free | |
1.5 mL | |
Non-looped | |
Screw Cap | |
20,000 x g |
Polypropylene | |
Polypropylene | |
O-ring | |
Conical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes