Learn More
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
Brand:  Axygen™ SCT-150-SS-C-S
 View more versions of this product
Product Code. 12719548
Description
Description
For collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Pre-sterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
Clear
1.5 mL
Polypropylene
O-ring
Sterile
Screw Cap
20,000 x g
RNase, DNase, pyrogen-free
Polypropylene
Clear
Non-looped
500/Pk.
No
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes > Clear; 1.5 mL; Self-standing; Yes