missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand: Axygen™ AM-2ML-RD
373.85 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Natural | |
Non-sterile | |
96 | |
10/Pk. | |
Round |
Silicone | |
Autoclavable | |
50/Cs. | |
Sealing Mat | |
96-well Microplate, 2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction