missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells
Minimize evaporation in wide range of temperatures in PCR and storage applications
401.18 € - 403.00 €
Specifications
Color | Natural |
---|---|
Material | Silicone |
Autoclavable | Autoclavable |
No. of Wells | 96 |
No. per Case | 50/Cs. |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
12557887
|
Axygen™
AM-2ML-RD |
Non-sterile |
403.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
12577887
|
Axygen™
AM-2ML-RD-S |
Sterile |
401.18 €
Pack of 50 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Natural | |
Autoclavable | |
50/Cs. | |
Sealing Mat | |
96-well Microplate, 2 mL |
Silicone | |
96 | |
10/Pk. | |
Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title