missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells

Minimize evaporation in wide range of temperatures in PCR and storage applications

Brand:  Axygen™ AM-2ML-RD-S

 View more versions of this product

Product Code. 12577887

  • 401.18 € / Pack of 50
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
Sterile
96
10/Pk.
Round
Silicone
Autoclavable
50/Cs.
Sealing Mat
96-well Microplate, 2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers