Learn More
Axygen™ Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Brand:  Axygen™ PCR-02CP-C
18.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12607385
  • 29.25 € / Pack of 125
Explore more special offers
Description
Description
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
125/Pk.
8-strips PCR tubes and 96-well PCR microplates
Polypropylene
Strip
Clear
0.2 mL
Polypropylene
Non-sterile
Domed Cap
8-Strip Domed PCR Tube Cap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Caps for PCR Tubes > Domed