missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Caps for PCR Tubes
GREENER_CHOICE

8-Strip domed or flat strip caps for Axygen PCR tubes

Brand:  Axygen™ PCR-02CP-C-100

28.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15409073

  • 39.40 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified RNase-DNase-free
0.2 mL
Non-sterile
Domed Cap
PCR Caps
Clear
8 strip PCR tubes and 96-well microplates
Polypropylene
1 x 8 Strip Caps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Caps for PCR Tubes > 0.2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.