Learn More
Axygen™ Microplate Sealing Film and Tapes
Offer great economy for disposable ease.
Brand: Axygen™ MF-300
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging
Specifications
Litto or Ethylene or Vinyl | |
5 in. | |
100/Cs. | |
127.2 x 79.5 mm | |
79.5 mm | |
Axygen PlateMax Semi-Automated plate sealer (HS-1120, HS-1230) | |
85 μm |
Non-sterile | |
127.2 mm | |
100/Pk. | |
3.12 in. | |
Designed for applications ranging from PCR and real-time PCR to storage | |
Heat Seal Film | |
-80°C to +104°C |
Product Suggestions
Customers who viewed this item also viewed