missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Offer great economy for disposable ease.

Brand:  Axygen™ PCR-AS-200

101.94 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 12577947

  • 191.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. 

Aluminum

  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum
5.74 in.
500/Cs.
146 x 79.2 mm
79.2 mm
Sealing Film
-80°C to +104°C
Non-sterile
146 mm
100/Pk.
3.11 in.
Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, ideal for light-sensitive samples
35 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Microplate Sealing Film and Tapes > Aluminum, Piercable; 64 μm; -80 °C - 104 °C; PCR Storage and Light Sensitive

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.