missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Multi Well High Profile Reagent Reservoirs
GREENER_CHOICE

Manufactured with virgin polypropylene for excellent chemical resistance

Brand:  Axygen™ RES-MW8-HP

201.43 EUR valid until 2024-12-31
Use promo code "22009" to get your promotional price.


 View more versions of this product

Product Code. 11330265

  • 349.46 € / Pack of 25
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI/SLAS, RNase/DNase-free, non-pyrogenic
80 mL
Non-sterile
25/Cs.
25/Cs.
8
Clear
Polypropylene
Reagent Reservoir
25/Pk.
10 mL
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers