Learn More
Axygen™ Multi Well High Profile Reagent Reservoirs

Manufactured with virgin polypropylene for excellent chemical resistance
349.46 € - 390.04 €
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Material | Polypropylene |
Type | Reagent Reservoir |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity & Availability | |||
11300265
![]() |
Axygen™
RES-MW12-HP-SI |
264 mL | Sterile | 12 | Individually Wrapped; 25/Cs. |
390.04 €
Pack of 25 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||
12537867
![]() |
Axygen™
RES-MW4-HP |
50 mL | Non-sterile | 4 | 25/Cs. |
360.09 €
Pack of 25 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
11311974
![]() |
Axygen™
RES-MW12-HP |
84 mL | Non-sterile | 12 | 25/Cs. |
351.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
11330265
![]() |
Axygen™
RES-MW8-HP |
80 mL | Non-sterile | 8 | 25/Cs. |
349.46 €
Pack of 25 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
11340265
![]() |
Axygen™
RES-MW8-HP-SI |
80 mL | Sterile | 8 | Individually Wrapped; 25/Cs. |
390.04 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
Polypropylene | |
Single-use |
Clear | |
Reagent Reservoir |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.