missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Multi Well Low Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
Brand: Axygen™ RES-MW8-LP
318.93 EUR valid until 2024-12-31
Use promo code "22009" to get your promotional price.
This item is not returnable.
View return policy
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
32 mL | |
Non-sterile | |
25/Cs. | |
25/Cs. | |
8 |
Clear | |
Polypropylene | |
Reagent Reservoir | |
25/Pk. | |
4 mL | |
Single-use |