missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Multi Well Low Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
349.46 € - 354.00 €
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Material | Polypropylene |
Type | Reagent Reservoir |
No. per Case | 25/Cs. |
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Packaging | Price | Quantity & Availability | ||
11310265
|
Axygen™
RES-MW12-LP |
84 mL | Non-sterile | 12 | 25/Cs. |
349.46 €
Pack of 25 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||
11320265
|
Axygen™
RES-MW12-LP-SI |
21 mL | Sterile | 12 | Individually Wrapped; 25/Cs. |
354.00 €
Pack of 25 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||
11350265
|
Axygen™
RES-MW8-LP |
32 mL | Non-sterile | 8 | 25/Cs. |
349.46 €
Pack of 25 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||
11360265
|
Axygen™
RES-MW8-LP-SI |
32 mL | Sterile | 8 | Individually Wrapped; 25/Cs. |
354.00 €
Pack of 25 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
Polypropylene | |
25/Cs. |
Clear | |
Reagent Reservoir | |
Single-use |