Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand: Axygen™ HS-400
This item is not returnable.
View return policy
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
Specifications
Plate Sealing | |
Polyester, Aluminum | |
100/Pk. |
PlateMax Plate Sealer | |
500/Cs. | |
Heat Sealing Film for Compound Storage |