missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
570.00 € - 598.79 €
Specifications
For Use With (Application) | Plate Sealing |
---|---|
For Use With (Equipment) | PlateMax Plate Sealer |
No. per Case | 500/Cs. |
No. per Pack | 100/Pk. |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12769618
|
Axygen™
HS-100-QPCR |
Heat Sealing Film for qPCR |
598.79 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12647765
|
Axygen™
HS-400 |
Heat Sealing Film for Compound Storage | N/A | ||||||
12627765
|
Axygen™
HS-200 |
Peelable Heat Sealing Film |
583.46 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12637765
|
Axygen™
HS-300 |
Pierceable Aluminum Heat Sealing Film |
583.46 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12779618
|
Axygen™
HS-150-QPCR |
Peelable Heat Sealing Film for qPCR |
598.79 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
Specifications
Plate Sealing | |
PlateMax Plate Sealer | |
500/Cs. | |
100/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title