Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE
Graduated tubes with conical bottom
Brand:  Axygen™ SCT-15ML-25-S
 View more versions of this product
Product Code. 12549067
  • 194.39 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
Not Autoclavable
Clear
For sample identification
25/Pk.
Sterile
Screw Cap
12,000 x g
HDPE
15 mL
Polypropylene
Screw cap
Bag
Yes
Cylindrical
Centrifuge Tube