Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Self-standing tubes
Brand:  Axygen™ ST-150-SS-C-S
 View more versions of this product
Product Code. 12739608
  • 598.91 € / Pack of 4000
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
Bag
Not Included
20,000 x g
1.5 mL
Polypropylene
Sterile
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms > Self-standing; Sterile; 1.5mL; Clear