missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Diamond Flex™ ESD Static Dissipative Workstation Mat
Notrax Diamond Flex™ ESD Static Dissipative Workstation Mat - The 18.4 mm thick platform elevates and insulates the worker from the hard ground while the bulleted domes underneath allow the mat to bend and 'flex' with every movement.
Brand: Notrax 548S3864BY
This item is not returnable.
View return policy
Description
- High quality anti-fatigue individual workstation mat for heavy-duty industrial environments
- Standard size mats for individual workstations are loose lay and require no installation
- The 184 mm thick platform elevates and insulates the worker from the hard ground while the bulleted domes underneath allow the mat to bend and ''flex'' with every movement
- Highly visible yellow safety ramps included on all sides to prevent tripping
- 100% nitrile rubber compound
Specifications
163 x 97 cm | |
5.3 ft. | |
Nitrile Rubber | |
97 cm |
Black, Yellow | |
163 cm | |
Industrial and Commercial Matting |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Notrax Diamond Flex™ ESD Static Dissipative Workstation Mat > Mat; Length: 163 cm; Width: 97 cm
Spot an opportunity for improvement?Share a Content Correction