missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates

For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-5ML-48-C-S
350.78 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
ANSI, SLAS 1-2004 | |
6 x 8 | |
Polypropylene | |
48 | |
Rectangular, Conical Bottom (V-well) | |
4000 x g | |
5/Pk. | |
-80°C |
Standard | |
Clear | |
5000 μL | |
Sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
25/Cs. | |
-112°F |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
AXY001 Catalogue GENERAL AXYGEN FS
File size: 2.2 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Storage Microplates > 48-well; 4.6 mL; Yes; Rectangular
Spot an opportunity for improvement?Share a Content Correction