missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates



For sample collection, in-vitro growth chambers and long term storage.
119.67 € - 374.10 €
Specifications
Format | Standard |
---|---|
Material | Polypropylene |
For Use With (Application) | Sample collection, in-vitro growth chambers and long term storage |
Temperature Range (English) | -112°F |
Temperature Range (Metric) | -80°C |
Product Code | Brand | Color | Sterility | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | No. of Wells | Volume (Metric) Well | Price | Quantity & Availability | |||||
12547907
![]() |
Axygen™
P-384-120SQ-C |
Clear | Non-sterile | 384 | 125 μL |
179.90 €
Pack of 50 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
12557907
![]() |
Axygen™
P-384-120SQ-C-S |
Clear | Sterile | 384 | 125 μL |
228.37 €
Pack of 50 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
12587907
![]() |
Axygen™
P-384-240SQ-C-S |
Clear | Sterile | 384 | 240 μL |
282.95 €
Pack of 50 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
11340115
![]() |
Axygen™
P-5ML-48-C-S |
Clear | Sterile | 48 | 5000 μL |
369.24 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11386074
![]() |
Axygen™
P-384-240SQ-C |
Clear | Non-sterile | 384 | 240 μL |
239.84 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
11330115
![]() |
Axygen™
P-5ML-48-C |
Clear | Non-sterile | 48 | 5000 μL |
257.78 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
12547917
|
Axygen™
P-96-450R-G |
Green | Non-sterile | 96 | 550 μL | N/A | ||||||
12587917
![]() |
Axygen™
P-96-450V-B |
Blue | Non-sterile | 96 | 550 μL |
137.03 €
Pack of 50 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
12587927
![]() ![]() |
Axygen™
P-DW-11-C-S |
Clear | Sterile | 96 | 1100 μL |
223.06 €
Pack of 50 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
12557837
![]() ![]() |
Axygen™
P-DW-10ML-24-C-S |
Clear | Sterile | 24 | 10,000 μL |
246.95 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||||
12577927
![]() ![]() |
Axygen™
P-DW-11-C |
Clear | Non-sterile | 96 | 1.1 mL |
228.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
12537837
![]() ![]() |
Axygen™
P-DW-10ML-24-C |
Clear | Non-sterile | 24 | 10,000 μL |
206.27 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||||
12597927
![]() ![]() |
Axygen™
P-DW-20-C |
Clear | Non-sterile | 96 | 1.8 mL |
262.00 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
11350135
![]() ![]() |
Axygen™
P-DW-20-C-S-IND |
Clear | Sterile | 96 | 2000 μL |
374.10 €
Pack of 30 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
|||||
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.Specifications
Standard | |
Sample collection, in-vitro growth chambers and long term storage | |
-80°C |
Polypropylene | |
-112°F |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Storage Microplates