missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
BIOPHARMAGREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-DW-20-C-S-IND

 View more versions of this product

Product Code. 11350135

  • 374.10 € / Pack of 30
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    ANSI, SLAS 1-2004
    8 x 12
    Clear
    Polypropylene
    96
    Round
    4000xG
    -80°C
    Standard
    Autoclavable
    96 Round Deep-Well Storage Microplate, Individually wrapped
    2000 μL
    Sterile
    Sample collection, in-vitro growth chambers and long term storage
    -112°F
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number24624052 Certificate TypeCertificate of Quality Date01/01/2025 Catalog NumberP-DW-20-C-S-IND
    Lot Number25024053 Certificate TypeCertificate of Quality Date14/12/2024 Catalog NumberP-DW-20-C-S-IND
    Lot Number19724102 Certificate TypeCertificate of Quality Date07/10/2024 Catalog NumberP-DW-20-C-S-IND
    Lot Number34123026 Certificate TypeCertificate of Quality Date15/05/2024 Catalog NumberP-DW-20-C-S-IND
    Lot Number34123026 Certificate TypeCertificate of Compliance Date26/03/2024 Catalog NumberP-DW-20-C-S-IND
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Axygen™ Storage Microplates > 96 round deep-well; Presterilized; Clear; 2.0mL, Individually wrapped

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.