missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-96-450R-G
This item is not returnable.
View return policy
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
ANSI, SLAS 1-2004 | |
8 x 12 | |
Green | |
Polypropylene | |
96 | |
Round | |
4000 x g | |
-80°C |
Standard | |
Autoclavable | |
96-well Round-bottom Storage Microplate | |
550 μL | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
-112°F |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Storage Microplates > 96-well assay; Round bottom plate Green; 500μL
Spot an opportunity for improvement?Share a Content Correction