Learn More
Tocris Bioscience™ AZD 7648
Potent and selective DNA-PK inhibitor
Brand:  Tocris Bioscience™ 7825/50
628.93 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18799076
  • 810.00 € / 50mg
Explore more special offers
Description
Description
AZD 7648 is a potent (IC50 = 0.6 nM in an enzyme assay) and selective DNA-PK inhibitor. It is selective for DNA-PK over a range of kinases including PI3Kα, PI3Kδ, PI3Kγ, ATM, ATR and mTOR. In combination with radiotherapy, it leads to tumor regression in fully immuno-competent mice models. It also enhances the activity of PARP inhibitor Olaparib (Cat. No. 7579). AZD 7648 is orally bioavailable.
Specifications
Specifications
7,9-Dihydro-7-methyl-2-[(7-methyl[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino]-9-(tetrahydro-2H-pyran-4-yl)-8H-purin-8-one
50 mg
C18H20N8O2
2230820-11-6
DNA-dependent Protein Kinase Inhibitors