missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 7648

Potent and selective DNA-PK inhibitor

210.00 € - 810.00 €

Specifications

Chemical Name or Material 7,9-Dihydro-7-methyl-2-[(7-methyl[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino]-9-(tetrahydro-2H-pyran-4-yl)-8H-purin-8-one
CAS 2230820-11-6
Target DNA-dependent Protein Kinase Inhibitors
Molecular Formula C18H20N8O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18779796
View Documents
Tocris Bioscience™
7825/10
10 mg
210.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18799076
View Documents
Tocris Bioscience™
7825/50
50 mg
810.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AZD 7648 is a potent (IC50 = 0.6 nM in an enzyme assay) and selective DNA-PK inhibitor. It is selective for DNA-PK over a range of kinases including PI3Kα, PI3Kδ, PI3Kγ, ATM, ATR and mTOR. In combination with radiotherapy, it leads to tumor regression in fully immuno-competent mice models. It also enhances the activity of PARP inhibitor Olaparib (Cat. No. 7579). AZD 7648 is orally bioavailable.
Specifications

Specifications

7,9-Dihydro-7-methyl-2-[(7-methyl[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino]-9-(tetrahydro-2H-pyran-4-yl)-8H-purin-8-one
2230820-11-6
DNA-dependent Protein Kinase Inhibitors
C18H20N8O2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documents

Documents

Product Certifications