Learn More
AZD-8055, MedChemExpress
MedChemExpress AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.
Brand: Medchem Express HY-10422
This item is not returnable.
View return policy
Description
AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.Specifications
AZD-8055 | |
Yellow | |
C25H31N5O4 | |
DMSO : 33.33 mg/mL (71.59 mM; Need ultrasonic) | |
465.54 | |
95.0% | |
Solid |
1009298-09-2 | |
Research | |
100 mg | |
OCC1=CC(C2=NC3=NC(N4CCOC[C@@H]4C)=NC(N5[C@@H](C)COCC5)=C3C=C2)=CC=C1OC | |
465.54 | |
Research | |
Cancer-Kinase/protease |