Learn More
AZD-8055, MedChemExpress
MedChemExpress AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.
91.80 € - 607.50 €
Specifications
Chemical Name or Material | AZD-8055 |
---|---|
CAS | 1009298-09-2 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C25H31N5O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16460317
|
Medchem Express
HY-10422 |
50 mg |
202.50 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16454976
|
Medchem Express
HY-10422 |
100 mg |
332.10 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16403968
|
Medchem Express
HY-10422 |
10 mg |
91.80 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440986
|
Medchem Express
HY-10422 |
200 mg |
607.50 €
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.Specifications
AZD-8055 | |
Yellow | |
C25H31N5O4 | |
OCC1=CC(C2=NC3=NC(N4CCOC[C@@H]4C)=NC(N5[C@@H](C)COCC5)=C3C=C2)=CC=C1OC | |
465.54 | |
Research | |
Cancer-Kinase/protease |
1009298-09-2 | |
Research | |
DMSO : 33.33 mg/mL (71.59 mM; Need ultrasonic) | |
465.54 | |
95.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.