missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 8055

Highly potent and selective mTOR inhibitor

147.00 € - 521.00 €

Specifications

Chemical Name or Material 5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol
CAS 1009298-09-2
Target mTOR Inhibitors
Molecular Formula C25H31N5O4
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30000027
View Documents
Tocris Bioscience™
7840/50
50 mg
521.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
30000023
View Documents
Tocris Bioscience™
7840/10
10 mg
147.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AZD 8055 is a highly potent, selective and ATP-competitive mTOR inhibitor (IC50 = 0.8 nM). Inhibits growth and induces autophagy in a range of cancer cell lines and xenografts, including NSCLC and glioblastoma (IC50 values are 20, 50 and 53 nM in H838, A549 and U87MG cell lines, respectively). Orally bioavailable.
Specifications

Specifications

5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol
mTOR Inhibitors
0.98
1009298-09-2
C25H31N5O4
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.