Learn More
Azlon™ Multi-lingual Safety Venting Wash Bottle
GREENER_CHOICE
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
39.25 € - 66.95 €
Specifications
Material LDPE
View More Specs
Products 21
Product Code Brand Capacity (Metric) Chemical Label Description Closure Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Chemical Label Description Closure Color Price Quantity & Availability  
12116230
View Documents
Azlon
WGW534VTML
250 mL I. M. S. Printed in English, French, German and Spanish Yellow
50.60 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
11373514
View Documents
Azlon
WGW531VTML
250 mL Acetone Printed in English, French, German and Spanish Red
39.25 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
12126230
View Documents
Azlon
WGW535VTML
250 mL Ethanol Printed in English, French, German and Spanish Orange
49.92 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
11383514
View Documents
Azlon
WGW532VTML
250 mL Methanol Printed in English, French, German and Spanish Green
41.95 €
Pack of 5
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
12196220
View Documents
Azlon
WGW533VTML
250 mL Isopropanol Printed in English, French, German and Spanish Blue
40.10 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
15307037
View Documents
Azlon
WGW537VTMLR
500 mL Acetone Region 3: Russian, Polish, German, English Red
54.00 €
Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
12327512
View Documents
Azlon
WGW537VTMLS
500 mL Acetone Region 2: Norwegian, Danish, Swedish, English Red
44.70 €
Pack of 5
Estimated Shipment: 18-07-2025
to see stock.
Add to basket
12357512
View Documents
Azlon
WGW541VTMLS
500 mL Ethanol Region 2: Norwegian, Danish, Swedish, English Orange
58.15 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
15387037
View Documents
Azlon
WGW541VTMLME
500 mL Ethanol Region 4: Arabic, French, English Orange
46.30 €
Pack of 5
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
15367037
View Documents
Azlon
WGW538VTMLME
500 mL Methanol Region 4: Arabic, French, English Green
46.45 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
15337037
View Documents
Azlon
WGW541VTMLR
500 mL Ethanol Region 3: Russian, Polish, German, English Orange
42.65 €
Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
11329643
GREENER_CHOICE
View Documents
Azlon
WGW537VTML
500 mL Acetone Printed in English, French, German and Spanish Red
66.45 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
15317037
missing translation for 'viewDocuments'
Azlon
WGW538VTMLR
500 mL Methanol Region 3: Russian, Polish, German, English Green
42.65 €
Pack of 5
Estimated Shipment: 14-04-2025
to see stock.
Aggiungi al carrello
15357037
missing translation for 'viewDocuments'
Azlon
WGW537VTMLME
500 mL Acetone Region 4: Arabic, French, English Red
45.50 €
Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Aggiungi al carrello
15377037
missing translation for 'viewDocuments'
Azlon
WGW539VTMLME
500 mL Isopropanol Region 4: Arabic, French, English Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12397512
missing translation for 'viewDocuments'
Azlon
WGW538VTMLS
500 mL Methanol Region 2: Norwegian, Danish, Swedish, English Green
42.65 €
Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Aggiungi al carrello
15327037
missing translation for 'viewDocuments'
Azlon
WGW539VTMLR
500 mL Isopropanol Region 3: Russian, Polish, German, English Blue
43.45 €
Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Aggiungi al carrello
12550456
missing translation for 'viewDocuments'
Azlon
WGW538VTML
500 mL Methanol Printed in English, French, German and Spanish Green
53.84 €
Pack of 5
In Stock
to see stock.
Edge
Aggiungi al carrello
11393514
missing translation for 'viewDocuments'
Azlon
WGW540VTML
500 mL I. M. S. Printed in English, French, German and Spanish Yellow
61.10 €
Pack of 5
In Stock
to see stock.
Edge
Aggiungi al carrello
11303524
GREENER_CHOICE
missing translation for 'viewDocuments'
Azlon
WGW539VTML
500 mL Isopropanol Printed in English, French, German and Spanish Blue
66.95 €
Pack of 5
In Stock
to see stock.
Edge
Aggiungi al carrello
11349643
GREENER_CHOICE
missing translation for 'viewDocuments'
Azlon
WGW541VTML
500 mL Ethanol Printed in English, French, German and Spanish Orange
42.95 €
Pack of 5
In Stock
to see stock.
Edge
Aggiungi al carrello
Descrizione
Descrizione
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
  • Multi-lingual wide neck wash bottles featuring the patented DripLok™ vapor-venting valve that prevents solvent drips
  • Indelibly printed in multiple languages (based on region)
  • Includes the Globally Harmonised Symbols, NFPA diamond, risk phrases, and safety information
  • Fitted with color coded one piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifica
Specifica
LDPE
Videos
SDS
Documenti
Documenti
Certificati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Azlon™ Multi-lingual Safety Venting Wash Bottle
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato