Learn More
Azlon™ LDPE DIY Printed Safety Wash Bottles
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
34.90 € - 67.80 €
Specifications
Color | Translucent |
---|---|
Diameter (Metric) Outer Neck | 38 mm |
Diameter (Metric) Inner Neck | 34 mm |
Includes | Closure, blank write-on panel, adhesive hazard signs |
Material | LDPE |
Product Code | Brand | Capacity (Metric) | Closure Type | Closure Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Type | Closure Color | Price | Quantity & Availability | |||
12540066
|
Azlon™
WGW001VTML |
250 mL | Vapor-venting | White |
60.65 €
Pack of 5 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
12510066
|
Azlon™
WGW001PML |
250 mL | Non-venting Cap | White |
34.90 €
Pack of 5 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
12520066
|
Azlon™
WGW002PML |
500 mL | Non-venting Cap | White |
61.10 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
12590056
|
Azlon™
WGW002VTML |
500 mL | Vapor-venting | White |
67.80 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
15307047
|
Azlon™
WGW004VTML |
500 mL | Vapor-venting | Assorted |
61.50 €
Each |
In Stock
Log in to see stock. |
|||
12500066
|
Azlon™
WGW020VTML |
1000 mL | Vapor-venting | White |
51.90 €
Pack of 5 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
12530066
|
Azlon™
WGW020PML |
1000 mL | Non-venting Cap | White |
43.75 €
Pack of 5 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||
Description
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
- Do-it-yourself wash bottles with blank write-on panel
- Indelibly printed in three languages (English, French and German)
- Supplied with adhesive hazard labels
Specifications
Translucent | |
34 mm | |
LDPE | |
Polypropylene |
38 mm | |
Closure, blank write-on panel, adhesive hazard signs | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.