missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polycarbonate Hinged Safety Shield

Protect yourself from potential hazards with a light weight but very strong safety shield. The Azlon™ polycarbonate hinged safety shield can be secured to a lab bench.

Brand:  Azlon SGT006

983.57 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10606855

  • 1180.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Excellent protection in potentially hazardous situations
  • Adjustable - can be used as a tight triangle or extended out to a straight line configuration
  • Incredibly strong and lightweight
  • Supplied with two pairs of plastic coated metal feet
  • Feet may be clamped or bolted onto the bench to give greater stability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hinged Safety Shield
61 cm
Clear
1EA
Polycarbonate
750 mm
For excellent protection in potentially hazardous situations
1EA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.