Learn More
Azlon™ Polystyrene Specimen Container with Polyethylene Lid
Store samples with maximum visibility. Azlon™ Polystyrene Specimen Containers with Polyethylene Lids contain snap-on lids that are very easy to use.
Brand:  Azlon BGG452P
 View more versions of this product
Product Code. 11363614
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Clear polystyrene containers with maximum sample visibility
  • Polyethylene airtight snap-on lids for quick easy use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Specimen Container
Polystyrene
150 mL
Clear
53 mm
10/Pk.
Yes
Non-sterile
Snap Cap
65 mm
Polyethylene lid